Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
Click to view available options
Type:
OxiTop™-IDS 1
OxiTop™-IDS 12
OxiTop™-i 2
OxiTop™-i 6
OxiTop™-i B
OxiTop™-i IS 12
OxiTop™-i IS 6
OxiTop™-i SET 2
Unit Size:
1 set
Each
Pack of 12
Description
- OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
- Measuring ranges up to 4000 mg/L BOD possible
- No need to convert data - direct input of sample volume
- Direct display of the BOD value on the measuring head screen
- Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
Specifications
Specifications
| Type | OxiTop™-i 2 |
| Display Type | Backlit Graphic LCD |
| Range | Measuring ranges up to 4000 mg/L BOD possible |
| Includes | Two OxiTop™-i measuring heads (1 x blue, 1 x grey) |