Learn More
Thermo Scientific™ Brilliance Candida Selective Supplement
Brilliance Candida is a chromogenic medium for the isolation and differentiation of Candida species.
Brand:  Thermo Scientific™ SR0231E
Product Code. 13285409
  • 191.00 EUR / Pack of 10
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Description
Brilliance Candida is a chromogenic medium for the isolation and differentiation of Candida species.
  • Add to to Brilliance Candida Agar Base, Part No. CM1002A and CM1002B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Brilliance Candida Selective Supplement
Selective Supplement
Each vial for 500mL medium
Freeze-dried
10/Pk.