Learn More
Thermo Scientific™ Oxoid™ Dermasel Selective Supplement
The Oxoid Dermasel Selective Supplement is used for Dermasel Medium, Part No. CM0539B.
Brand:  Thermo Scientific™ SR0075E
Product Code. 11434159
  • 164.00 EUR / Pack of 10
Explore more special offers
Description
Description
Oxoid Dermasel Selective Supplement is used for Dermasel Medium, Part No. CM0539B.
Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dermasel Selctv Supp.
Selective Supplement
Each vial for 500mL medium
Freeze-dried
10/Pk.