Learn More
Thermo Scientific™ Oxoid™ Dermasel Selective Supplement
The Oxoid Dermasel Selective Supplement is used for Dermasel Medium, Part No. CM0539B.
Brand: Thermo Scientific™ SR0075E
This item is not returnable.
View return policy
Description
Oxoid Dermasel Selective Supplement is used for Dermasel Medium, Part No. CM0539B.
Each vial supplements 500mL of mediumSpecifications
Dermasel Selctv Supp. | |
Selective Supplement | |
Each vial for 500mL medium |
Freeze-dried | |
10/Pk. |