Learn More
Thermo Scientific™ Oxoid™ Streptococcus Supplement
Product Code. 11484189
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 11484189
Brand: Thermo Scientific™ SR0126E
Oxoid Streptococcus Supplement is used for the isolation of Streptococci.
Oxoid Streptococcus Supplement is used for the isolation of Streptococci.
  • Add to Columbia Blood Agar Base, Part No. CM0331B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Streptococcus Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Streptococcus species
compliance-icons
Product Identifier
  • Streptococcus Selective Supplement
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : Colistin, sulfate (salt), Oxolinic acid