missing translation for 'onlineSavingsMsg'
Learn More

P3H3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 EUR - 410.00 EUR

Specifications

Antigen P3H3
Concentration 0.16 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16835474
View Documents
Proteintech
16023-1-AP-20UL
20 μL
145.00 EUR
20µL
Please to purchase this item. Need a web account? Register with us today!
16825474
View Documents
Proteintech
16023-1-AP-150UL
150 μL
410.00 EUR
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene belongs to the leprecan family of proteoglycans, which function as collagen prolyl hydroxylases that are required for proper collagen biosynthesis, folding and assembly. This protein, like other family members, is thought to reside in the endoplasmic reticulum. Epigenetic inactivation of this gene is associated with breast and other cancers, suggesting that it may function as a tumor suppressor.
Specifications

Specifications

P3H3
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Mouse, Rat, Human
Q8CG70, Q8IVL6
10536, 14789, 297595
P3H3 Fusion Protein Ag8850
Primary
-20°C
P3H3
0.16 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
GRCB, HSU47926, leprecan like 2, Leprecan like protein 2, LEPREL2, P3H3, Prolyl 3 hydroxylase 3, Protein B
P3H3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications