missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p97/VCP Rabbit anti-Human, Mouse, Rat, Clone: 3P8T3, Novus Biologicals™
Rabbit Monoclonal Antibody
191.14 EUR - 483.79 EUR
Specifications
Antigen | p97/VCP |
---|---|
Clone | 3P8T3 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18374352
|
Bio-Techne
NBP3-15478-20UL |
20 μg |
191.14 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18331627
|
Bio-Techne
NBP3-15478-100UL |
100 μg |
483.79 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p97/VCP Monoclonal antibody specifically detects p97/VCP in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
p97/VCP | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cellular Markers, DNA Repair, Membrane Trafficking and Chaperones, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7415 | |
IgG | |
Affinity purified |
3P8T3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human p97/VCP (P55072). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |