Learn More
PAβN dihydrochloride, MedChemExpress
MedChemExpress PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.
Brand: Medchem Express HY-101444A
This item is not returnable.
View return policy
Description
PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.Specifications
PAβN dihydrochloride | |
100929-99-5 | |
25 mg | |
C25H32Cl2N6O2 | |
MC-207,110 dihydrochloride Phe-Arg-β-naphthylamide dihydrochloride | |
O=C([C@H](CCCNC(N)=N)NC([C@H](CC1=CC=CC=C1)N)=O)NC2=CC(C=CC=C3)=C3C=C2.Cl.Cl | |
519.47 | |
Research |
Solid | |
White | |
Research | |
COVID-19-immunoregulation | |
DMSO : 83.33 mg/mL (160.41 mM; Need ultrasonic) ∣H2O : 14.29 mg/mL (27.51 mM; Need ultrasonic) | |
519.47 | |
98.51% |