missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PADI4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17373-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at ′Arg-8′ and/or ′Arg-17′ and histone H4 at ′Arg-3′, which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at ′Arg-2142′, which favors its interaction with NCOA2/GRIP1.Specifications
PADI4 | |
Polyclonal | |
Unconjugated | |
PADI4 | |
HL 60 PAD, PAD, PAD4, PADI4, PADI5, PDI4, PDI5, Peptidylarginine deiminase IV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18602, 23569, 29512 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88807, Q9UM07, Q9Z183 | |
Padi4 | |
PADI4 Fusion Protein Ag11068 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |