Learn More
PAL International™ PAL TX Surface Disinfectant Wipes Pocket Probe Canister
A small pocket size portable dispensing canister for convenient disinfection of probes and utensils
Brand:  PAL International™ W65231T
Product Code. 15874477
  • 179.00 EUR / Pack of 48
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Description
  • Perfect for laboratory testing in food factories – both at production line side and in the lab itself
  • Can also be easily stored for use in food retail outlets where temp probes are used regularly
  • Pal TX Surface Disinfectant wipes are food safe and proven to kill most microorganisms such as bacteria, yeast & viruses
  • Pal TX Surface Disinfectant Wipes clean and disinfect without using Quats, PHMB or alcohol
Specifications
Specifications
Pocket Canister
70 x 130 mm
Disinfection of probes and utensils
70 Wipes/Canister, 48 Canisters/Pk.