Learn More
Hach Pacific Pty Ltd PAN Indicator Solution, 0.1%
For determination of low level manganese by the PAN method.
55.30 EUR
Description
For determination of low level manganese by the PAN method. Hach method 8149. Range: 0.006 to 0.700 mg/L Mn.
- Specifications
- Method Name: PAN
- Platform: Solution
Specifications
0.006 to 0.700 mg/L | |
Solution |