Learn More
Memmert Pass-through Oven UF750TS TwinDISPLAY, 749L, 20-250°C, Forced Convection
Brand:  Memmert UF750TS
14740.59 EUR valid until 2025-12-20
BEST PRICE on Promo! Use promo code "24121" to get your promotional price.
Pass-through Oven UF750TS TwinDISPLAY, 749L, 20-250°C, Forced Convection
Product Code. 16318037
  • ONLINE EXCLUSIVE PRICE
    18060.00 EUR
    17511.00 EUR / Each
    Save 549.00 EUR
    3% Off
Only null left
Estimated Shipment: 29-01-2026