Learn More
ratiolab™ Pasteur-Plast Pipets
Click to view available options
:
Pasteur-Plast Pipets; Graduated: No; Length (Metric): 104 mm; Sterility: Non-sterile; Volume (Metric): 1 mL
Description
- Soft, elastic bulb is unbreakable, thus reducing the risk of glass injury
- Burr-free orifice for neat, clean and precise dispensing
- Only externally graduated Pasteur-plast pipets with clearly visible embossed graduations
Specifications
Specifications
| Material | LDPE |
| Sterility | Non-sterile |
| Drops/mL | 50 Drops |
| Graduated | No |
| Quantity | 400 Pipets/Box |
| Temperature (Metric) Operating | ≤70°C |
| Color | Clear |
| Volume (Metric) | 1 mL |
| Length (Metric) | 104 mm |
| Type | Pasteur Pipet |
| Show More |