Learn More
Emma Safety Footwear Patrick XD Safety Shoes
Product Code. 15968961
Click to view available options
Size:
37
38
39
40
41
42
45
46
48
Unit Size:
1 pair
Product Code. 15968961
Brand: Emma Safety Footwear MM73186642
Only null left
Add to basket
Add to basket
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design Patrick XD
Material Leather
Size 42