missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCF11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23540-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to the 173 kDa human PCF11 protein.
Component of pre-mRNA cleavage complex II.Specifications
PCF11 | |
Polyclonal | |
Unconjugated | |
PCF11 | |
KIAA0824, PCF11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51585 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94913 | |
PCF11 | |
PCF11 Fusion Protein Ag20263 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |