missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCYT1A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
168.46 EUR - 343.40 EUR
Specifications
Antigen | PCYT1A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331725
|
Bio-Techne
NBP3-15502-20UL |
20 μg |
168.46 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18344664
|
Bio-Techne
NBP3-15502-100UL |
100 μg |
343.40 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCYT1A Polyclonal antibody specifically detects PCYT1A in Human, Mouse samples. It is validated for Western BlotSpecifications
PCYT1A | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
CCT A, CCT-alpha, choline-phosphate cytidylyltransferase A, CT, CT A, CTA, CTP:phosphocholine cytidylyltransferase A, CTPCTphosphate cytidylyltransferase 1, choline, alpha isoform, EC 2.7.7, EC 2.7.7.15, PCYT1CCTA, phosphate cytidylyltransferase 1, choline, alpha, Phosphorylcholine transferase A | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PCYT1A (NP_001299602.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Neuroscience, Signal Transduction | |
PBS, 50% glycerol, pH7.3 | |
5130 | |
IgG | |
Affinity purified |