Learn More
asecos™ PE IBC Station with Galvanised Grid
Shop All asecos Products
Click to view available options
Width (Metric):
1370.00 mm
2530.00 mm
Unit Size:
Each
Description
- Complete corrosion protection
- Highly resistant against oil, acid, alkali, etc.
- Easy and safe in-house transport of the empty sump by forklift truck
Specifications
Specifications
| Color | Blue |
| Capacity (English) | 264.2 gal. |
| Capacity (Metric) | 1000 L |
| Material | Polyethylene |
| Type | IBC Spill Pallet |
| Length (English) | 53.94 in. |
| Length (Metric) | 1370.00 mm |
| Width (English) | 53.94 in. |
| Width (Metric) | 1370.00 mm |
| Height (English) | 27.17 in. |
| Show More |