Learn More
Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint
Click to view available options
Diameter (Metric) Outer:
27 mm
33 mm
42 mm
51 mm
64 mm
85 mm
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
Socket Joint:
14/23 NS
19/26 NS
29/32 NS
Unit Size:
Pack of 10
Description
- Acc. to DIN 12383
- With inscription label
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Borosilicate Glass, Duran Glass |
| Socket Joint | 14/23 NS |
| Diameter (Metric) Outer | 51 mm |