missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Pectinase from Aspergillus niger
Merk: MP Biomedicals™ 0219897905
This item is not returnable.
View return policy
Beschrijving
Pectinase is a macerating enzyme comprised of pectintranseliminase, polygalacturonase, pectinesterase, xylanase, hemicellulases and cellulases.
- Lyophilized
- Activity: >1000 u/gm

Specificatie
>1000 U/g | |
Used in plant protoplast preparation to digest cell wall prior to organelle isolation | |
Aspergillus niger | |
Pectinase |
Beige | |
4°C | |
5 kU | |
Lyophilized Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction