missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Penicillin-Streptomycin (20,000 IU Pen/mL, 20,000 μg Strep/mL)
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
- The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria
- This solution contains 17,000 to 21,000 units/mL of penicillin and 18.0 to 23.0 mg/mL of streptomycin
Specifications
Specifications
| Content And Storage | Stored at -20°C |
| Antibiotic Spectrum | Prevent bacterial contamination of cell cultures, Gram-Positive and Gram-Negative Bacteria |
| Concentration | 20,000 IU Pen/mL, 20,000 μg Strep/mL |
| Form | Liquid |
| Product Type | Antibiotic |
| Sterility | Sterile |
| For Use With (Application) | Cell Culture |
| Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction