missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorophenylhydrazine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Descrizione
Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.
Solubility
Soluble in methanol.
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.
Solubility
Soluble in methanol.
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Specifica
Specifica
| Melting Point | 73°C to 78°C |
| Quantity | 5 g |
| Beilstein | 747800 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 198.09 |
| Percent Purity | 97% |
| Chemical Name or Material | Pentafluorophenylhydrazine |
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto