missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene), Thermo Scientific Chemicals

47.10 EUR - 150.75 EUR

Chemical Identifiers

CAS 2070-70-4
Molecular Formula C6F12
Molecular Weight (g/mol) 300.05
MDL Number MFCD00153253
InChI Key SAPOZTRFWJZUFT-UHFFFAOYSA-N
Synonym 2z-1,1,1,2,3,4,5,5,5-nonafluoro-4-trifluoromethyl pent-2-ene
PubChem CID 11012007
SMILES FC(=C(F)C(F)(C(F)(F)F)C(F)(F)F)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313786
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L16836.14
25 g
47.10 EUR
25g
Please to purchase this item. Need a web account? Register with us today!
11323786
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L16836.22
100 g
150.75 EUR
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2070-70-4
300.05
SAPOZTRFWJZUFT-UHFFFAOYSA-N
11012007
C6F12
MFCD00153253
2z-1,1,1,2,3,4,5,5,5-nonafluoro-4-trifluoromethyl pent-2-ene
FC(=C(F)C(F)(C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

2070-70-4
(CF3)2CFCF=CFCF3
MFCD00153253
SAPOZTRFWJZUFT-UHFFFAOYSA-N
1,1,1,2,3,4,5,5,5-nonafluoro-4-(trifluoromethyl)pent-2-ene
11012007
≥90%
Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1]
C6F12
<1.3000
2z-1,1,1,2,3,4,5,5,5-nonafluoro-4-trifluoromethyl pent-2-ene
FC(=C(F)C(F)(C(F)(F)F)C(F)(F)F)C(F)(F)F
300.05
300.05
contains 5 to 10% perfluoro(2-methyl-2-pentene)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only