missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Perform™ Classic Concentrate Mucasol

Product Code. p-200104824 Shop All Brand Products
Click to view available options
Size:
2 L
5 L
Unit Size:
2 L
5 L
This item is not returnable. View return policy

Product Code. 17991302

Brand: BRAND™ 44103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths.

  • Liquid, alkaline high-performance concentrate for immersion and ultrasonic baths.
  • Universal cleaner for laboratory equipment and instruments made of glass, porcelain, plastic, rubber and metal.
  • Items are cleaned in about 10 to 30 minutes, without mechanical help. Perform™ classic concentrate mucasol replaces chromosulfuric acid, is free from chlorine and caustic alkalis, is non-toxic, nonaggressive and biodegradable.
  • Prolonged exposure of zinc and non-anodized aluminium surfaces should be avoided.
TRUSTED_SUSTAINABILITY

Specifications

Color-coded No
Form Concentrated Liquid
Scent Original
Sterility Non-sterile
Appearance Light Yellow
Autoclavable Not Autoclavable
Color Translucent Yellow
Disposable Limited-use
Filtered Filtered
Flammable No
Size 2 L
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Perform Concentrate Mucasol Liquid Detergent
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : tripotassium orthophosphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.