Learn More
Labcon™ PerformR™ 15 mL Polystyrene Centrifuge Tubes
Click to view available options
Sterility:
Non-sterile
Sterile
Quantity:
1000 Tubes/Pk.
125 Tubes/Bag, 4 Bags /Cs.
25 Tubes/Low Waste Fiber Rack, 2 Racks/Pk., 10 Pk./Cs.
50 Tubes/Bag, 10 Bags/Cs.
Unit Size:
Pack of 1000
Pack of 125
Pack of 500
Description
- Conical centrifuge tube
- ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
- Caps include a foamed low density polyethylene (LDPE) liner
- Not autoclavable or freezable
- Max RCF: 3,000 xg
- Made in USA
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP |
| Closure Material | Polyethylene, LDPE Foam |
| Closure Color | Purple |
| Color | Natural |
| Capacity (Metric) | 15 mL |
| Disposable | Single-use |
| Material | Polystyrene |
| For Use With | Centrifuges, Rotators, Shakers |
| No. per Pack | 25/Pk. |
| Packaging | 25 Tubes/Reusable Rack, 2 Racks/Pk., 10 Pk./Cs. |
| Show More |