Learn More
Watson-Marlow 120 Series Drive with Pumphead
Click to view available options
Flow Rate:
0.0001 to 36 mL/min.
0.001 to 120 mL/min.
0.001 to 170 mL/min.
0.002 to 190 mL/min.
0.003 to 54 mL/min.
Pump Head:
3-channel Pump Head
Flip-top Pump Head
Quick-change Pump Head
Single-channel Pump Head
Twin-channel Pump Head
Unit Size:
Each
Description
- The 120 series is the perfect pump for maximising yield in fermentation and cell culture applications. Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimise your process. The 120 series pump is perfectly suited to single-use systems. Error-free tube change takes only seconds.
- Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate. With an exceptional 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials. A keypad lock secures your pumping parameters for consistent and repeatable performance.
- 120R single channel, two roller pumphead for continuous tubing
Pumphead options available as standard (others available on request):
Specifications
Specifications
| Control Type | Digital Variable Speed |
| Display Type | LED |
| Enclosure Rating | IP31 |
| Flow Rate | 0.001 to 120 mL/min. |
| Maximum Pressure | 30 psi |
| Motor Speed | 0.01 to 200 rpm |
| External Control | Manual |
| Model | 120S/D1 |
| Pump Head | Single-channel Pump Head |
| Voltage | 90/264 V |