missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Peroxide-Cured Silicone Pump Tubing

Reduce the risk of particulate entrapment with smooth inner surface

Brand:  Ismatec™ 07625-32

Product Code. 15155952

  • 167.00 EUR / 15m
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Specifications

Specifications

Microbore Tubing
1.3 mm
Plastic tubing and other Microbore tubing
1 Each
230°C
Autoclavable
Silicone
-50°C
32
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.