Learn More
Haldenwanger™ Pestle
Product Code. 10245164
Click to view available options
Diameter (Metric) Outer:
24mm
28mm
30mm
42mm
45mm
48mm
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. 10245164
Brand: Haldenwanger™ L 56/4/GLASIERT
Currently Unavailable: We have no information when this product will be available.
View alternative products
Pestle
Grinding surfaces are glazed.
Specifications
Diameter (Metric) Outer 45mm
Product Type Pestle
Length (Metric) 180mm
Type Pestle