missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pet1 Antibody (FEV/7311) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23997
This item is not returnable.
View return policy
Description
Pet1 Monoclonal antibody specifically detects Pet1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Pet1 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry-Paraffin: 1-2 μg/mL | |
| FEV (ETS oncogene family), FEV (fifth Ewing variant), Fifth Ewing variant protein, HSRNAFEV, PC12 ETS domain-containing transcription factor 1, PC12 ETS factor 1, Pet-1, PET1, protein FEV | |
| Recombinant fragment (around aa1-200) of human Pet1 protein (exact sequence is proprietary) | |
| 100 μg | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 54738 | |
| Store at -20°C to -80°C. Avoid freeze/thaw cycles. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| FEV/7311 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction