missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Petroleum Ether

Product Code. 16696153
Click to view available options
Quantity:
7 L
18 L
200 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Petroleum Ether, Puriss., low boiling point hydrogen treated naphtha, meets analytical specification of DAB, 40°C to 60°C (bp min. 75%).

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 64742-49-0
MDL Number MFCD00081849
Synonym Petroleum benzin
IUPAC Name Petroleum ether

Specifications

pH Not applicable
Melting Point No data available
CAS Min % >50.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.65 g/cm3 (20°C)
Boiling Point 40°C to 60°C (1.013 hPa)
Quantity 7 L
Flash Point <-30°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss.
Vapor Pressure No data available
UN Number UN3295
Viscosity No data available
Grade Pharmacopeia
Chemical Name or Material Petroleum ether
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.