Learn More
PF-4878691, MedChemExpress
Product Code. 16482818
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16482818
Brand: Medchem Express HY100176
MedChemExpress PF-4878691 (3M-852A) is a potent, orally active, and selective Toll-like receptor 7 (TLR7) agonist modelled to dissociate its antiviral and inflammatory activities.
PF-4878691 (3M-852A) is a potent, orally active, and selective Toll-like receptor 7 (TLR7) agonist modelled to dissociate its antiviral and inflammatory activities.
Specifications
Chemical Name or Material PF-4878691
Physical Form Solid
CAS 532959-63-0
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C17H23N5O2S
For Use With (Application) COVID-19-anti-virus
Synonym 3M-852A
Solubility Information DMSO : 19.23 mg/mL (53.20 mM; Need ultrasonic)
SMILES CS(=O)(NCCCCN1C(CC)=NC2=C1C3=CC=CC=C3N=C2N)=O
Molecular Weight (g/mol) 361.46
Formula Weight 361.46
Percent Purity 98.97%
Grade Research