missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN5 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22337AF532
This item is not returnable.
View return policy
Description
PFDN5 Polyclonal antibody specifically detects PFDN5 in Human, Mouse samples. It is validated for Western BlotSpecifications
PFDN5 | |
Polyclonal | |
Western Blot | |
c-myc binding protein, C-Myc-binding protein Mm-1, MM-1, MM1MGC71907, Myc modulator 1, myc modulator-1, PFD5MGC5329, prefoldin 5, prefoldin subunit 5 | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Prefoldin Subunit 5 using the numbering given in entry NP_002615.2 (GeneID 5204). | |
0.1 mL | |
Signal Transduction | |
5204 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |