Learn More
pH 7.413 DIN 19266 Buffer Solution at 25°C, Reagecon
BIOPHARMA
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand:  Reagecon 107413
Product Code. 11862643
Description
Description
  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
Accuracy ± 0.010 pH units
Liquid
HDPE Bottle GL45
7.413 ±0.010
500 mL