Learn More
SI Analytics™ pH Combination Electrode with 1m Fixed Cable
Ensure high measurement accuracy, stability, and long service life while maintaining high adaptability to your measurement tasks. SI Analytics™ pH Combination Electrode with 1m Fixed Cable is robust, durable, and easy to clean.
Brand:  SI Analytics™ 285105451
 View more versions of this product
Product Code. 10151601
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Proven high-end electrodes for demanding measurement tasks
  • Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
  • Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
  • Each pH combination electrode comes with individual serial number and quality certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BNC
A-Glass
pH Combination Electrode
Sphere
-5°C to +100°C
1 m
Platinum Diaphragm
0 to 14
Silamid™
N 52 BNC
120 mm
12 mm