Learn More
SI Analytics™ pH Glass Electrode
SI Analytics™ pH Glass Electrode is constructed of a plastic shaft with a screw plug head.
Brand:  SI Analytics™ 285110208
Product Code. 10461073
  • 242.00 EUR / Each
Estimated Shipment: 24-10-2024
to see stock.
Explore more special offers
Description
  • Ag / AgCl reference
  • Zeropoint pH = 7.0
  • N-glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
N-glass
Plastic Shaft
N 2981
0°C to 80°C
12 mm
Screw Plug Head
0 to 14
Sphere
120 mm