Learn More
Fisherbrand™ pH Indicator Paper Rolls
Shop All Fisherbrand Products
Click to view available options
pH Range:
0 to 10
0 to 11
1 to 14
5.5 to 9
6.4 to 8
Format:
Refill
Roll
Unit Size:
Each
Pack of 4
Description
- Filter paper strips soaked with one or several coloured indicators of pH
- Economical selection of indicator papers cover the entire range of pH 0 to 14
- Available in a variety of formats to meet individual needs
- Reference chart shows colour changes in steps of pH2
Specifications
Specifications
| pH Range | 1 to 14 |
| Format | Refill |
| Includes | Color scale printed on cover |
| Quantity | 3/Pk. |
| Length (Metric) | 5 m |