Learn More
Masterflex™ PharMed Opaque Tubing
Click to view available options
Diameter (Metric) Inner:
12.8 mm
Unit Size:
Each
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 12.8 mm |
| Diameter (Metric) Outer | 19.1 mm |
| Overall Length (Metric) | 7.62 m |
| Thickness (Metric) | 3.2 mm |
| Min. Operating Temperature (Metric) | -59°C |
| Packaging Quantity | 1 Each |
| Sterility | Sterile |
| Color | Opaque |
| Maximum Pressure | 11 psi |
| Show More |