Learn More
MP Biomedicals™ Phillips and Hart Salt Mixture
Shop All MP Biomedicals Products
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
Description
- Salt Mixture
- For research animal diet
- Powder
Specifications
Specifications
| Product Type | Animal Diet |
| For Use With (Application) | Research animal diet |
| Quantity | 2 kg |
| Storage Requirements | Room Temperature |