missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-cGAS (Ser291) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117261
This item is not returnable.
View return policy
Description
Positive Samples: 293T Immunogen sequence: PGSPA.
cGAS (Cyclic GMP-AMP synthase) is a nucleotidyltransferase that catalyzes the formation of cyclic GMP-AMP (cGAMP) from ATP and GTP and plays a key role in innate immunity. Catalysis involves both the formation of 2', 5' phosphodiester linkage at the GpA step and the formation of 3', 5' phosphodiester linkage at the ApG step. This produces c[G(2',5')pA(3'5')p] responses. cGAS also binds cytosolic DNA directly, leading to the activation and synthesis of cGAMP. Then, a second messenger binds to and activates TMEM173/STING thereby triggering type-I interferon production. cGAS is also involved in innate immune sensorship of infection, detection of DNA from bacteria, response to cellular stress, DNA damage, and regulation of cellular senescence.
Specifications
| Phospho-cGAS (Ser291) | |
| Polyclonal | |
| Unconjugated | |
| CGAS | |
| C6orf150; h-cGAS; MB21D1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 115004, 214763 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.76 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q8C6L5, Q8N884 | |
| CGAS | |
| Synthetic Peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction