missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-EphA2 (Ser897) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105547
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of EPHA2 only when phosphorylated at Ser897.
EPHA2 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. EPHA2 is overexpressed in many types of human cancers. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
Specifications
| Phospho-EphA2 (Ser897) | |
| Polyclonal | |
| Unconjugated | |
| EPHA2 | |
| ARCC2; AW545284; CTPA; CTPP1; CTRCT6; Eck; EPH receptor A2; Epha2; ephrin receptor EphA2; Ephrin type-A receptor 2; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; I79_019726; Myk2; Sek2; Sek-2; soluble EPHA2 variant 1; tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor SEK-2 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 13836, 1969, 366492 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P29317, Q03145 | |
| EPHA2 | |
| A synthesized peptide derived from human EPHA2(Accession P29317), corresponding to amino acid residues around phosphorylated Ser897. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction