missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus lump, Red, Puratronic™, 99.999+% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 7723-14-0
Molecular Formula P
Molecular Weight (g/mol) 30.97
MDL Number MFCD00133771
InChI Key OAICVXFJPJFONN-UHFFFAOYSA-N
Synonym red, acmc-20 ajw2
PubChem CID 5462309
ChEBI CHEBI:30207
IUPAC Name phosphorus
SMILES [P]
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7723-14-0
30.97
OAICVXFJPJFONN-UHFFFAOYSA-N
5462309
phosphorus
P
MFCD00133771
red, acmc-20 ajw2
CHEBI:30207
[P]

Specifications

7723-14-0
Liquid
P
1338
Miscible with water
[P]
30.97
CHEBI:30207
Powder or Lump
1.701 g/cm3 (14.195 lb./gal)
Brown to Red
Puratronic™, 99.999+% (metals basis)
MFCD00133771
red, acmc-20 ajw2
OAICVXFJPJFONN-UHFFFAOYSA-N
phosphorus
5462309
300°C
7100 hPa (5325 mm Hg)
Phosphorus lump, red
Videos
Safety and Handling

Safety and Handling

Hazard statements

H228 Flammable solid

Precautionary statements

  • P210 Keep away from heat/sparks/open fl ames/hot surfaces. No smoking.
  • P280 Wear protective gloves/protective clothing/eye protection/face protection.
  • P240 Ground/bond container and receiving equipment.
  • P241 Use explosion-pr oof electrical/ventila ting/lighting/equipment.
  • P370+P378 In case of fire: Use for ex tinction: CO2, powder or water spray.

Danger

missing translation for 'dotInformation' : 4.1 Flammable solids

missing translation for 'einecsNumber' : 231-768-7

missing translation for 'rtecsNumber' : TH3495000

missing translation for 'tsca' : YEs

SDS
Documents

Documents

Product Certifications