Learn More
uvex Phynomic WET Water-Resistant Gloves with Aqua-Polymer Foam Coating
Click to view available options
Size:
8
9
10
11
12
Description
- Water-repellent safety gloves with an aqua-polymer foam coating for use in outdoor areas
- Breathable coating is durable to provide a high degree of mechanical abrasion resistance
- Enhanced grip capabilities in damp and wet areas
- Tactile feel when assembling parts
- Ideal for precision work, maintenance, and repairs
Specifications
Specifications
Construction | Seamless Knit |
Coating Material | Aqua-Polymer Foam |
Color | Blue, Black |
Exterior Finish | Smooth |
Hand | Ambidextrous |
Material | Spandex, Polyamide |
Product Type | Mechanical Protection Gloves |
Size | 9 |
Style | Full-hand Mitt |
EN388 Standard | 3131X |