Learn More
Sartorius Picus™ NxT Electronic Pipettes, 8 Channels
BIOPHARMAGREENER_CHOICE
Product Code. 15320635 Shop All Sartorius Products
Click to view available options
Volume (Metric):
0.2 to 10 μL
10 to 300 μL
5 to 120 μL
50 to 1200 μL
Color:
Gray
Orange
Purple
Yellow
missing translation for 'unitSize'
Each
Product Code. 15320635
missing translation for 'mfr': Sartorius LH745321
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to ease user's workload and to protect from repetitive strain injury (RSI)
Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.
  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management
  • Volume range: 0.2 μL to 1200 μL
  • Delivered without USB AC adapter (available separately)
Applications:
PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture
Certifications
CE Marked
Services
2 year
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Number of Channels 8
Product Line Picus
Type Electronic Pipette
Volume (Metric) 0.2 to 10 μL