Learn More
Thermo Scientific™ Pierce™ Ninhydrin (2,2-dihydroxy-1H-indene-1,3(2H)-dione)
Product Code. 11876794 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. 11876794
Brand: Thermo Scientific™ 21003BID
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Label, detect and measure peptides, proteins and amino acids by HPLC (liquid chromatography) using this amine-reactive, colored reagent (Amax 440nm).
Thermo Scientific™ Pierce Ninhydrin is a classic, amine-reactive, colorimetric detection reagent for protein detection and amino acid analysis.
Highlights:
  • Used in amino acid chromatography
  • Offers superb color response and low blank
Ordering Alerts:
Indefinitely stable. No refrigeration required. Keep bottle tightly sealed. Avoid exposure to direct sunlight and ammonia.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ninhydrin
Chemical Reactivity Amine
Product Line Pierce
Content And Storage Store in a segregated and approved area. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Label or Dye Other Label or Dye
Detection Method Colorimetric
Quantity 500 g