missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sulfo-SIAB (sulfosuccinimidyl (4-iodoacetyl)aminobenzoate)
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
missing translation for 'unitSize'
50mg
Description
Thermo Scientific™ Pierce Sulfo-SIAB a water-soluble, mid-length (10.6 Å) crosslinker for amine-to-sulfhydryl conjugation via sulfo-N-hydroxysuccinimide (sulfo-NHS) ester and iodoacetyl reactive groups.
Highlights:
- Water-soluble
- Reactive groups: sulfo-NHS ester and iodoacetyl
- Reactive toward: amino and sulfhydryl groups
Specifications
Specifications
| Chemical Reactivity | Amine-Sulfhydryl |
| Cleavable | No |
| Description | Sulfo-SIAB |
| Molecular Weight (g/mol) | 504.19 |
| PEGylated | No |
| Spacer Arm Length | 10.6 Å |
| Content And Storage | Upon receipt store at -20°C protected from light. |
| Cell Permeability | No |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction