Learn More
Thermo Scientific™ Piko PCR Plate, 24-well, clear
Product Code. 15276207
Click to view available options
Quantity:
200 plates
50 plates
Unit Size:
Pack of 200
Pack of 50
Product Code. 15276207
Brand: Thermo Scientific™ SPL024050
Only null left
Add to basket
Add to basket
Thermo Scientific™ Piko PCR Plates are modular, ultra-thin wall PCR plates for Piko* Thermal Cyclers.
Thermo Scientific™ Piko™ PCR Plates are modular, ultra-thin wall PCR plates for Piko Thermal Cyclers.
>Highlights
• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 24-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 96-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions
Applications
• PCR
Related Products
Piko PCR Plate, 24-well, white
Piko PCR Plate, 24-well, clear
Plate Frame for 24-well Piko PCR Plates, white
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certifications/Compliance Certified RNase/DNase-free
Packaging 50 Plate
Product Line Piko
Color Clear
Product Type PCR Microplate
For Use With (Equipment) Piko Thermal Cycler, PikoReal Real-Time PCR System
Material Polypropylene
No. of Wells 24
Quantity 50 plates
Reaction Speed Low profile