Learn More
OHAUS™ Pioneer™ Precision Balance
Product Code. p-7217313 Shop All Ohaus Products
Click to view available options
Capacity:
620 g
6200 g
Readability:
0.001 g
0.01 g
Model:
PX6202
PX6202/E
PX623
PX623/E
Unit Size:
1 unit
Product Code. 17278770
Brand: OHAUS™ 30619385
Affordable balance to achieve reliable results
  • Offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and education settings at an economical price point
  • Featuring cast metal lower housing, sub-pan and stainless steel weighing pan
    • Durably constructed for versatile, long-term use
  • Second line display for additional information/guidance and USB/RS232 connectivity for easy communication
    • draft shield models features static removal (ESR) bar for convenient grounding
Key Features:
  • User-selectable environmental filters and brightness settings
  • Auto-tare
  • Auto-dim
  • User-selectable span calibration points
  • Software lockout and reset menu
  • User selectable communication settings and data print options
  • User-definable project and user IDs
TRUSTED_SUSTAINABILITY
Specifications
Connection Type RS232 and USB device (included), GLP/GMP data output with real-time clock
Capacity 6200 g
Display Type Backlit LCD
Readability 0.01 g
For Use With (Application) Basic weighing, Parts counting, Percent weighing, Animal weighing, Density determination
Linearity 0.02 g
Material Metal base, Plastic top housing, Removable stainless steel pan, Glass draft shield
Model PX6202
Repeatability 0.01 g
Stabilization Time 1 sec.
Temperature Range (English) 50 to 86 °F, Operability guaranteed between 41 to 104 °F
Temperature Range (Metric) 10 to 30 °C, Operability guaranteed between 5 to 40 °C
Type Analytical and precision balances
Weighing Modes Gram, Kilogram, Ounce, Pound, Carat, Pennyweight, Ounce Troy, Grain, Newton, Hong Kong Tael, Singapore Tael, Taiwan Tael, Momme, Tical, Mesghal, Tola, Baht, Custom
Dimensions (L x W x H) 321 x 209 x 98 mm
Length (Metric) 321 mm
Width (Metric) 209 mm
Height (Metric) 98 mm
Diameter (Metric) Outer Pan 180 mm
Weight (English) 7.7 lb.
Weight (Metric) 3.5 kg