missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperaquine tetraphosphate tetrahydrate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.
Specifications
Specifications
| Chemical Name or Material | Piperaquine tetraphosphate tetrahydrate |
| Physical Form | Solid |
| CAS | 915967-82-7 |
| Color | White |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C29H32Cl2N6.4H3O4P.4H2O |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | H2O : 6 mg/mL (6.00 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (5.00 mM; Need ultrasonic and warming) |
| SMILES | ClC1=CC=C2C(N3CCN(CCCN4CCN(C5=CC=NC6=CC(Cl)=CC=C56)CC4)CC3)=CC=NC2=C1.OP(O)(O)=O.[4].[4 H2O] |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction