Learn More
Piperaquine tetraphosphate tetrahydrate, MedChemExpress
Product Code. 16431138
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16431138
Brand: Medchem Express HYB1896B
MedChemExpress Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.
Piperaquine tetraphosphate tetrahydrate is a bisquinoline antimalarial agent. Piperaquine tetraphosphate tetrahydrate can be used in antimalarial research in combination with Artemisinin.
Specifications
Chemical Name or Material Piperaquine tetraphosphate tetrahydrate
Physical Form Solid
CAS 915967-82-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C29H32Cl2N6.4H3O4P.4H2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 6 mg/mL (6.00 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (5.00 mM; Need ultrasonic and warming)
SMILES ClC1=CC=C2C(N3CCN(CCCN4CCN(C5=CC=NC6=CC(Cl)=CC=C56)CC4)CC3)=CC=NC2=C1.OP(O)(O)=O.[4].[4 H2O]
Molecular Weight (g/mol) 999.6
Formula Weight 999.6
Percent Purity 98.0%
Grade Research