missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Pipetus™ Standard Pipet Filler
Provides the best prerequisites for working in potentially explosive environments, with toxic reagents or in clean room technology through the separation of the electric pump and hand-held unit. Hirschmann Instruments™ Pipetus™ Standard Pipet Filler enables highly accurate pipetting through fine dispensing valves with valve pins made of ceramic compound. A safety valve prevents the penetration of liquid into the interior of the unit.
Brand: Hirschmann™ 9903000
This item is not returnable.
View return policy
Description
Provides the best prerequisites for working in potentially explosive environments, with toxic reagents or in clean room technology through the separation of the electric pump and hand-held unit. Further sophisticated product details guarantee precision, reliability and a long service life. Highly accurate pipetting is enabled through fine dispensing valves with valve pins made of ceramic compound. A safety valve prevents the penetration of liquid into the interior of the unit. The standard PTFE diaphragm filter provided with 0.2μm pore size ensures reliable protection against overfilling. The sophisticated ergonomic shape of the pipetus™ standard aids non-fatiguing work.- Highly accurate pipetting enabled through fine dispensing valves with valve pins made of ceramic compound
- The seat of all pipets is sealed absolutely tight with a silicone adapter
- Extremely light hand unit with external pump
- Hydrophobic PTFE diaphragm filter with 0.2 μm pore size
- For measuring, volumetric and Pasteur pipettes, from 0.1 to 200mL
Specifications
PTFE | |
0.2 μm | |
accurate pipetting | |
0.1 to 200mL | |
Euro | |
230 V |
Standard | |
Pipette Filler | |
Measuring, volumetric, and Pasteur pipets | |
Pipet holder with safety valve and filter | |
Hand component: 130 g, pump, 290 g |