Learn More
Microspec™ Plastic Inoculation Loops
Click to view available options
Volume:
1 μL
5 μL
10 μL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 250
Description
- Includes 250 Plastic Inoculation Loops
- Aseptic Manufactured or Ethylene Oxide Sterilized
- 200 mm length
Specifications
Specifications
| Disposable | Single-use |
| Volume | 10 μL |
| Sterility | Non-sterile |
| Color | Blue |
| Length (Metric) Handle | 200 mm |
| Quantity | 250 |
| Packaging Type | Microseal Pouch |
| Autoclavable | Not Autoclavable |