Learn More
KD Scientific Plastic Syringe
Product Code. 15154204
Click to view available options
Volume (Metric):
1 mL
3 mL
5 mL
10 mL
20 mL
30 mL
Connection Type:
Luer Lock
Luer Slip
Tuberculin Luer Slip
Unit Size:
Pack of 25
Product Code. 15154204
Brand: KD Scientific 780857
Economical and disposable, available in sizes ranging from 1 cc to 50 cc, Luer Slip (LS) or Luer Lock (LL) tip
  • A new line of disposable plastic sterile syringes for all scientific applications
  • Norm-ject syringes are the ideal solution for any situation
  • Their unique two-part system is latex free and contains no silicone lubricant or rubber
Specifications
Material Polyethylene (Plunger)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Syringe
Disposable Single-use
Volume (Metric) 3 mL
For Use With (Equipment) KD Scientific Syringe Pumps
Connection Type Luer Slip
Graduated Yes